Assay Details
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756656.3 | XP_002756702.1 | 3 - 4 | 1710 | 96 | |
XM_009010180.2 | XP_009008428.1 | 4 - 5 | 1142 | 96 | ||
XM_009010182.2 | XP_009008430.1 | 3 - 4 | 1710 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
