Assay Details
Gene Symbol: LYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758913.3 | XP_002758959.1 | 3 - 4 | 469 | 61 | |
XM_008982662.2 | XP_008980910.1 | 3 - 4 | 383 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
