Assay Details
| Gene Symbol: LOC100398815 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985994.1 | XP_008984242.1 | 3 - 4 | 388 | 84 | |
| XM_017967026.1 | XP_017822515.1 | 3 - 4 | 1170 | 84 | ||
More Information
Set Membership: |
|

LOC100398815
Genomic Map