Assay Details
| Gene Symbol: C5H17orf51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971685.1 | XP_017827174.1 | 2 - 3 | 479 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5H17orf51| Gene Symbol: C5H17orf51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971685.1 | XP_017827174.1 | 2 - 3 | 479 | 63 | |
Set Membership: |
|