Assay Details
Gene Symbol: PTPN20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017978928.1 | XP_017834417.1 | 1 - 2 | 29 | 67 | |
XM_017978931.1 | XP_017834420.1 | 1 - 2 | 103 | 67 | ||
XM_017978932.1 | XP_017834421.1 | 1 - 2 | 29 | 67 | ||
XM_017978933.1 | XP_017834422.1 | 1 - 2 | 29 | 67 | ||
XM_017978935.1 | XP_017834424.1 | 1 - 2 | 111 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017978931.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
