Assay Details
| Gene Symbol: PTPN20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978928.1 | XP_017834417.1 | 7 - 8 | 914 | 117 | |
| XM_017978929.1 | XP_017834418.1 | 6 - 7 | 830 | 117 | ||
| XM_017978930.1 | XP_017834419.1 | 4 - 5 | 692 | 117 | ||
| XM_017978931.1 | XP_017834420.1 | 7 - 8 | 992 | 117 | ||
More Information
Set Membership: |
|

PTPN20
Genomic Map