Assay Details
Gene Symbol: OTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747465.3 | XP_002747511.1 | 1 - 2 | 694 | 126 | |
XM_017971973.1 | XP_017827462.1 | 1 - 2 | 699 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() |
