Assay Details
Gene Symbol: SYNGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762321.2 | XP_002762367.1 | 3 - 4 | 626 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
