Assay Details
Gene Symbol: KCNA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003733517.3 | XP_003733565.1 | 1 | 1522 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
