Assay Details
Gene Symbol: KDELC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754367.2 | XP_002754413.1 | 2 - 3 | 450 | 70 | |
XM_017977520.1 | XP_017833009.1 | 2 - 3 | 450 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
