Assay Details
| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977431.1 | XP_017832920.1 | 7 - 8 | 1981 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SAMD4A| Gene Symbol: SAMD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977431.1 | XP_017832920.1 | 7 - 8 | 1981 | 59 | |
Set Membership: |
|