Assay Details
Gene Symbol: CHST14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753545.4 | XP_002753591.1 | 1 | 321 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
