Assay Details
| Gene Symbol: ACSM2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755957.4 | XP_002756003.1 | 3 - 4 | 1227 | 78 | |
| XM_009009265.2 | XP_009007513.1 | 4 - 5 | 1135 | 78 | ||
| XM_009009267.2 | XP_009007515.1 | 3 - 4 | 1227 | 78 | ||
| XM_009009269.2 | XP_009007517.1 | 3 - 4 | 1227 | 78 | ||
More Information
Set Membership: |
|

ACSM2B
Genomic Map