Assay Details
| Gene Symbol: C10H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754318.3 | XP_002754364.1 | 2 - 3 | 517 | 88 | |
| XM_009006645.2 | XP_009004893.1 | 2 - 3 | 497 | 88 | ||
| XM_017977365.1 | XP_017832854.1 | 2 - 3 | 209 | 88 | ||
More Information
Set Membership: |
|

C10H14orf180
Genomic Map