Assay Details
Gene Symbol: ITPRIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008980059.2 | XP_008978307.1 | 1 | 721 | 83 | |
XM_008980060.2 | XP_008978308.1 | 2 | 477 | 83 | ||
XM_017964061.1 | XP_017819550.1 | 2 | 480 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
