Assay Details
| Gene Symbol: BCKDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988070.2 | XP_008986318.1 | 3 - 4 | 639 | 65 | |
| XM_008988071.2 | XP_008986319.1 | 3 - 4 | 387 | 65 | ||
More Information
Set Membership: |
|

BCKDHA
Genomic Map