Assay Details
Gene Symbol: LOC100390425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017963436.1 | XP_017818925.1 | 10 - 11 | 962 | 104 | |
XM_017963437.1 | XP_017818926.1 | 15 - 16 | 1754 | 104 |

More Information
Set Membership: |
![]() ![]() |
