Assay Details
Gene Symbol: LOC100390425 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017963436.1 | XP_017818925.1 | 6 - 7 | 615 | 69 | |
XM_017963437.1 | XP_017818926.1 | 11 - 12 | 1407 | 69 |

More Information
Set Membership: |
![]() ![]() |
