Assay Details
Gene Symbol: LOC100394535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756370.2 | XP_002756416.1 | 3 - 4 | 251 | 89 | |
XM_009009931.1 | XP_009008179.1 | 3 - 4 | 276 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
