Assay Details
Gene Symbol: MBNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008989898.2 | XP_008988146.1 | 2 - 3 | 240 | 63 | |
XM_008989900.2 | XP_008988148.1 | 2 - 3 | 301 | 63 | ||
XM_008989901.2 | XP_008988149.1 | 2 - 3 | 345 | 63 | ||
XM_008989904.2 | XP_008988152.1 | 2 - 3 | 266 | 63 | ||
XM_008989905.2 | XP_008988153.1 | 2 - 3 | 188 | 63 | ||
XM_017968539.1 | XP_017824028.1 | 2 - 3 | 645 | 63 | ||
XM_017968540.1 | XP_017824029.1 | 2 - 3 | 345 | 63 | ||
XM_017968541.1 | XP_017824030.1 | 2 - 3 | 424 | 63 | ||
XM_017968542.1 | XP_017824031.1 | 2 - 3 | 345 | 63 | ||
XM_017968543.1 | XP_017824032.1 | 2 - 3 | 255 | 63 | ||
XM_017968544.1 | XP_017824033.1 | 3 - 4 | 431 | 63 | ||
XM_017968545.1 | XP_017824034.1 | 2 - 3 | 385 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
