Assay Details
| Gene Symbol: PRIMPOL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008992395.2 | XP_008990643.1 | 14 - 15 | 1914 | 77 | |
| XM_008992398.2 | XP_008990646.1 | 12 - 13 | 1509 | 77 | ||
| XM_008992399.2 | XP_008990647.1 | 12 - 13 | 1662 | 77 | ||
| XM_008992400.2 | XP_008990648.1 | 13 - 14 | 1626 | 77 | ||
| XM_017970111.1 | XP_017825600.1 | 13 - 14 | 1822 | 77 | ||
| XM_017970112.1 | XP_017825601.1 | 13 - 14 | 1834 | 77 | ||
| XM_017970113.1 | XP_017825602.1 | 11 - 12 | 1629 | 77 | ||
More Information
Set Membership: |
|

PRIMPOL
Genomic Map