Assay Details
| Gene Symbol: KRT71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003926.1 | XP_009002174.1 | 5 - 6 | 1164 | 71 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRT71| Gene Symbol: KRT71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003926.1 | XP_009002174.1 | 5 - 6 | 1164 | 71 | |