Assay Details
Gene Symbol: TNFRSF10D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017963690.1 | XP_017819179.1 | 1 - 2 | 273 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
