Assay Details
Gene Symbol: TNFRSF10D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017963690.1 | XP_017819179.1 | 4 - 5 | 598 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
