Assay Details
| Gene Symbol: USP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968215.1 | XP_017823704.1 | 1 | 2624 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP29| Gene Symbol: USP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968215.1 | XP_017823704.1 | 1 | 2624 | 101 | |
Set Membership: |
|