Assay Details
Gene Symbol: ODF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753357.3 | XP_002753403.1 | 2 - 3 | 1036 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
