Assay Details
| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733349.3 | XP_003733397.3 | 10 - 11 | 1704 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA21| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733349.3 | XP_003733397.3 | 10 - 11 | 1704 | 58 | |
Set Membership: |
|