Assay Details
| Gene Symbol: C8H7orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002168.1 | XP_009000416.1 | 3 - 4 | 161 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C8H7orf65| Gene Symbol: C8H7orf65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009002168.1 | XP_009000416.1 | 3 - 4 | 161 | 86 | |
Set Membership: |
|