Assay Details
| Gene Symbol: DNAAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977084.1 | XP_017832573.1 | 2 - 3 | 2435 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAAF2| Gene Symbol: DNAAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977084.1 | XP_017832573.1 | 2 - 3 | 2435 | 113 | |
Set Membership: |
|