Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756144.4 | XP_002756190.2 | 5 - 6 | 2106 | 80 | |
XM_009009599.2 | XP_009007847.1 | 6 - 7 | 2099 | 80 | ||
XM_009009601.2 | XP_009007849.1 | 6 - 7 | 2099 | 80 | ||
XM_009009603.2 | XP_009007851.1 | 5 - 6 | 575 | 80 | ||
XM_017978940.1 | XP_017834429.1 | 7 - 8 | 2175 | 80 | ||
XM_017978941.1 | XP_017834430.1 | 6 - 7 | 2099 | 80 |
