Assay Details
| Gene Symbol: BRICD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017978637.1 | XP_017834126.1 | 2 - 3 | 1032 | 54 | |
| XM_017978638.1 | XP_017834127.1 | 2 - 3 | 1032 | 54 | ||
More Information
Set Membership: |
|

BRICD5
Genomic Map