Assay Details
| Gene Symbol: SNRPD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743599.4 | XP_002743645.1 | 3 - 4 | 347 | 67 | |
| XM_009007861.2 | XP_009006109.1 | 2 - 3 | 1178 | 67 | ||
More Information
Set Membership: |
|

SNRPD3
Genomic Map