Assay Details
| Gene Symbol: EDARADD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760884.3 | XP_002760930.1 | 5 - 6 | 1326 | 68 | |
| XM_008985773.2 | XP_008984021.1 | 5 - 6 | 860 | 68 | ||
More Information
Set Membership: |
|

EDARADD
Genomic Map