Assay Details
Gene Symbol: C19H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008985741.2 | XP_008983989.1 | 5 - 6 | 642 | 84 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C19H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008985741.2 | XP_008983989.1 | 5 - 6 | 642 | 84 |