Assay Details
Gene Symbol: LOC100394842 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008997150.2 | XP_008995398.1 | 2 - 3 | 386 | 75 | |
XM_017972558.1 | XP_017828047.1 | 3 - 4 | 1621 | 75 | ||
XM_017972559.1 | XP_017828048.1 | 3 - 4 | 1625 | 75 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
