Assay Details
| Gene Symbol: NSMCE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749060.3 | XP_002749106.1 | 1 | 1516 | 163 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NSMCE3| Gene Symbol: NSMCE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749060.3 | XP_002749106.1 | 1 | 1516 | 163 | |
Set Membership: |
|