Assay Details
Gene Symbol: LOC100389294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749383.3 | XP_002749429.1 | 12 - 13 | 2363 | 95 | |
XM_002749384.3 | XP_002749430.1 | 12 - 13 | 2303 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
