Assay Details
| Gene Symbol: TSGA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003064.2 | XP_009001312.1 | 6 - 7 | 987 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSGA13| Gene Symbol: TSGA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003064.2 | XP_009001312.1 | 6 - 7 | 987 | 67 | |
Set Membership: |
|