Assay Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003735300.3 | XP_003735348.1 | 1 - 2 | 247 | 77 | |
XM_008985917.2 | XP_008984165.1 | 1 - 2 | 247 | 77 | ||
XM_008985918.2 | XP_008984166.1 | 1 - 2 | 258 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
