Assay Details
| Gene Symbol: SLCO6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965366.1 | XP_017820855.1 | 4 - 5 | 891 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLCO6A1| Gene Symbol: SLCO6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965366.1 | XP_017820855.1 | 4 - 5 | 891 | 106 | |
Set Membership: |
|