Assay Details
Gene Symbol: TAS2R7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752181.2 | XP_002752227.2 | 1 | 812 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
