Assay Details
Gene Symbol: GUCA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758826.3 | XP_002758872.1 | 2 - 3 | 463 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GUCA1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758826.3 | XP_002758872.1 | 2 - 3 | 463 | 70 |
Set Membership: |
![]() ![]() ![]() |