Assay Details
| Gene Symbol: TXNDC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001215.2 | XP_008999463.1 | 2 - 3 | 462 | 81 | |
| XM_017974738.1 | XP_017830227.1 | 2 - 3 | 537 | 81 | ||
More Information
Set Membership: |
|

TXNDC12
Genomic Map