Assay Details
| Gene Symbol: KLRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752334.3 | XP_002752380.1 | 3 - 4 | 563 | 83 | |
| XM_017975945.1 | XP_017831434.1 | 3 - 4 | 572 | 83 | ||
More Information
Set Membership: |
|

KLRG1
Genomic Map