Assay Details
Gene Symbol: TMEM207 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758195.3 | XP_002758241.1 | 2 - 3 | 226 | 86 | |
XM_008981390.1 | XP_008979638.1 | 2 - 3 | 226 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
