Assay Details
Gene Symbol: CDK5R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003733242.2 | XP_003733290.2 | 1 | 817 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
