Assay Details
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008983129.2 | XP_008981377.1 | 7 - 8 | 1278 | 70 | |
XM_017965548.1 | XP_017821037.1 | 7 - 8 | 1038 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
