Assay Details
Gene Symbol: SLAMF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008984736.2 | XP_008982984.1 | 1 - 2 | 327 | 71 | |
XM_008984737.2 | XP_008982985.1 | 1 - 2 | 339 | 71 | ||
XM_008984738.2 | XP_008982986.1 | 1 - 2 | 326 | 71 | ||
XM_008984739.2 | XP_008982987.1 | 1 - 2 | 339 | 71 | ||
XM_017966403.1 | XP_017821892.1 | 1 - 2 | 379 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
