Assay Details
| Gene Symbol: C9H12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003637.2 | XP_009001885.1 | 9 - 10 | 1439 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9H12orf40| Gene Symbol: C9H12orf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003637.2 | XP_009001885.1 | 9 - 10 | 1439 | 90 | |