Assay Details
Gene Symbol: FHL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971266.1 | XP_017826755.1 | 5 - 6 | 1020 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
